Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Slug Antibody (OTI1A6), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274229PE
Description
Slug Monoclonal specifically detects Slug in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
Slug | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
SNAI2 | |
Full length recombinant protein of human Slug (NP_003059) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
OTI1A6 | |
Flow Cytometry | |
MGC10182, Neural crest transcription factor Slug, Protein snail homolog 2, slug homolog, zinc finger protein (chicken), SLUGH, SLUGzinc finger protein, snail 2, snail homolog 2 (Drosophila), WS2D, zinc finger protein SNAI2 | |
Mouse | |
Affinity Purified | |
Neuronal Cell Markers, Neuronal Stem Cell Markers, Phospho Specific | |
6591.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction