Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Slug Antibody (OTI2B11), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274235MFV450
Description
Slug Monoclonal antibody specifically detects Slug in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
Slug | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI2B11 | |
Flow Cytometry | |
MGC10182, Neural crest transcription factor Slug, Protein snail homolog 2, slug homolog, zinc finger protein (chicken), SLUGH, SLUGzinc finger protein, snail 2, snail homolog 2 (Drosophila), WS2D, zinc finger protein SNAI2 | |
Full length human recombinant protein of human SNAI2 (NP_003059) produced in E.coli. | |
0.1 mL | |
Neuronal Cell Markers, Phospho Specific | |
6591 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction