Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLUG Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5120209
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120209 Supplier Thermo Scientific Supplier No. PA5120209
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse brain, Rat lung, Rat spleen. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: ELDTHTVIIS PYLYESYSMP VIPQPEILSS GAYSPITVWT TAAPFHAQLP NGLSPLSGYS SSLGRVSPPP PSDTSSKDHS GSESPISDEE ERLQSKLSDP

Slug, a member of the Snail family of C2H2-type zinc finger transcription factors, was initially identified to be involved in epithelial-mesenchymal transitions as well as the formation of the neural tube during vertebrate embryogenesis. Like Snail, Slug transcription can be induced by growth factors such as FGF, BMP, and TGF-beta. Once expressed, Slug will bind E-box regions of promoters and repress transcription of genes such as E-cadherin and Claudin-1. More recently, its expression in breast, esophogeal, and colorectal carcinomas has been correlated with poor prognosis for survival. Furthermore, Slug can protect hemopoietic progenitor cells from radiation-induced apoptosis by repressing the p53-mediated transcription of Puma, a BH3-only antagonist of the anti-apoptotic members of the Bcl-2 family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLUG
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SNAI2
Gene Accession No. O08954, O43623, P97469
Gene Alias MGC10182; neural crest transcription factor Slug; Protein snail homolog 2; SLUG; slug (chicken homolog), zinc finger protein; Slug, chicken homolog; Slugh; SLUGH1; Snai2; snail family transcriptional repressor 2; snail family zinc finger 2; snail homolog 2; Snail2; snail-like protein 2; WS2D; Zinc finger protein SNAI2; zinc finger transcription factor snail 2
Gene Symbols SNAI2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 21-120 of human Slug (NP_0030591)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20583, 25554, 6591
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.