Learn More
Invitrogen™ SLUG Polyclonal Antibody

Description
In Western blot, this antibody detects a band at ~30kDa in 293 cell lysate. Rabbit anti Slug antibody detects Slug, a member of the Snail family of zinc finger transcription factors, whose expression is induced by FGF, BMP, and TGF-beta.
Specifications
Specifications
| Antigen | SLUG |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SNAI2 |
| Gene Accession No. | O43623, P97469 |
| Gene Alias | MGC10182; neural crest transcription factor Slug; Protein snail homolog 2; SLUG; slug (chicken homolog), zinc finger protein; Slug, chicken homolog; Slugh; SLUGH1; Snai2; snail family transcriptional repressor 2; snail family zinc finger 2; snail homolog 2; Snail2; snail-like protein 2; WS2D; Zinc finger protein SNAI2; zinc finger transcription factor snail 2 |
| Gene Symbols | SNAI2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.