Learn More
Invitrogen™ SLURP1 Monoclonal Antibody (569317)

Description
In direct ELISAs, no cross-reactivity with recombinant mouse SLURP1 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | SLURP1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 569317 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | SLURP1 |
| Gene Accession No. | P55000 |
| Gene Alias | anti-neoplastic urinary protein; ANUP; ARS; ARS component B; ARS(component B)-81/S; ArsB; LY6LS; lymphocyte antigen 6-like secreted; MDM; secreted LY6/PLAUR domain containing 1; secreted Ly6/uPAR related protein 1; secreted Ly-6/uPAR-related protein 1; SLURP1; SLURP-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.