Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLX1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561410
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561410 Supplier Invitrogen™ Supplier No. PA561410
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGCLLRAHVI CLAEEFLQEE PGQLLPLEGQ CPCCEKSLLW GDLIWLCQMD TEKEVEDSEL EEAHWTDLLE T Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.

SLX1 encodes a protein that is an important regulator of genome stability. The protein represents the catalytic subunit of the SLX1-SLX4 structure-specific endonuclease, which can resolve DNA secondary structures that are formed during repair and recombination processes. Two identical copies of this gene are located on the p arm of chromosome 16 due to a segmental duplication. SLZ1A represents the more centromeric copy and SLZ1B represents the more telomeric copy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLX1A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLX1A
Gene Accession No. Q9BQ83
Gene Alias SLX1A; SLX1B
Gene Symbols SLX1A, SLX1B
Host Species Rabbit
Immunogen Recombinant protein fragment of Human SLX1A. Recombinant protein control fragment (Product #RP-104420).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 548593, 79008
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.