Learn More
SLX4IP/C20orf94 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304995A

Description
The recommended shelf life for this product is 1 year from date of receipt.
SLX4IP/C20orf94 interacts with SLX4/BTBD12.Specifications
SLX4IP/C20orf94 | |
Polyclonal | |
Unconjugated | |
SLX4IP | |
bA204H22.1, bA254M13.1, C20orf94, chromosome 20 open reading frame 94, dJ1099D15.3, SLX4-interacting protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
128710 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q5VYV7 | |
SLX4IP | |
Between 175 and 225 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.