Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SM 324405
GREENER_CHOICESDP

Catalog No. 544550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-455-0 50 mg
54-451-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 54-455-0 Supplier Tocris Bioscience™ Supplier No. 5445/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent TLR7 agonist

SM 324405 is a potent TLR7 agonist (EC50 = 50 nM). Exhibits selectivity for TLR7 over TLR8. Induces IFN-α and -γ expression in human PBMCs and mouse splenocytes, and inhibits IL-5 production in human PBMCs in vitro. Rapidly metabolized to acid metabolite (t½ = 2.6 min in human plasma).

Specifications

Chemical Name or Material Methyl 3-[(6-amino-2-butoxy-7,8-dihydro-8-oxo-9H-purin-9-yl)methyl]benzeneacetate
CAS 677773-91-0
Quantity 50 mg
Target Toll-like Receptor Agonists
Molecular Formula C19H23N5O4
Formula Weight 385.42
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.