Learn More
Sm-D3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | Sm-D3 |
---|---|
Applications | Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-6509
![]() |
Bethyl Laboratories, Inc
A303953AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
The protein belongs to the small nuclear ribonucleoprotein core protein family. It is required for pre-mRNA splicing and small nuclear ribonucleoprotein biogenesis.Specifications
Sm-D3 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
SMD3, Sm-D3, small nuclear ribonucleoprotein Sm D3, small nuclear ribonucleoprotein polypeptide D3, snRNP core protein D3 | |
SNRPD3 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P62318 | |
6634 | |
Between 1 and 50 | |
Primary | |
4° C | |
SNRPD3 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.