Learn More
Cincinnati Surgical Swann Morton Sterile Stainless Steel Blades and Scalpels
SDP
Catalog No. 19200244
Click to view available options
Size:
# 10
# 10A
# 11
# 12
# 15
# 20
# 21
# 22
# 23
# 25A
Product Type:
Blade
Scalpel
Scalpel Blade
Catalog No. 19-200-244
Supplier: Cincinnati Surgical 01SM25A
Only null left
Add to Cart
Add to Cart
For use in wide variety of medical applications, including Dermaplaning
  • Blades packaged with VCI Paper to protect blade from corrosion
  • Scalpels individually packaged in transparent thermos formed tray
  • 200% visual inspection
  • Strongest/Smallest foil package available
  • Narrow grinding angle resulting in increased initial keenness
  • High surface finish
  • Cellophane overwrap
  • Extended range
Specifications
Blade Material Stainless Steel
Product Type Blade
Size # 25A
Sterility Sterile
Packaging Type Packaged with VCI Paper to protect Blade from Corrosion
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.