Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SM047 Antibody (SM047), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

$181.90 - $487.50

Specifications

Antigen SM047
Clone SM047
Dilution Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:100-1:1000
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP197921SS
SDP
View Documents
Novus Biologicals
NBP197921SS
0.025 mL
Each for $181.90
Only null left
Add to Cart
 
NBP197921
SDP
View Documents
Novus Biologicals
NBP197921
0.1 mL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

SM047 Monoclonal specifically detects SM047 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

SM047
Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:100-1:1000
Monoclonal
Ascites
RUO
Human
anti-SMO47
IgM
Unpurified
Glycocalyx of human ovarian carcinoma cells.
SM047
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Ovarian Carcinoma Cell Markers
Ascites with 0.1% Sodium Azide
Cell membranes from human ovarian serous cystadenocarcinmoma and cells from human ovarian cancer cell line UWOV1.
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.