Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SM22 Alpha Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984563
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4563 Supplier Bioss Supplier No. BS2178R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAGLN is a transformation and shape-change sensitive actin cross-linking/gelling protein found in fibroblasts and smooth muscle. Its expression is down-regulated in many cell lines, and this down-regulation may be an early and sensitive marker for the onset of transformation. A functional role of this protein is unclear.

Specifications

Antigen SM22 Alpha
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TAGLN
Gene Accession No. Q01995
Gene Alias 22 kDa actin-binding protein; 25 kDa F-actin-binding protein; Actin-associated protein p27; BOS_14359; DKFZp686B01212; DKFZp686P11128; Protein WS3-10; Sm22; Sm22a; Sm22alpha; SM22-alpha; SMCC; smooth muscle 22 protein; Smooth muscle protein 22-alpha; Tagln; TAGLN1; Transgelin; transgelin variant 2; Ws310; WS3-10
Gene Symbols TAGLN
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SM22 Alpha.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6876
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.