Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smac/DIABLO Mouse anti-Human, Unconjugated, Clone: 2D12, Southern Biotech™
SDP

Catalog No. OB1107001 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB1107001 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB1107001 Supplier Southern Biotechnology Associates Supplier No. 1107001

Mouse Monoclonal Antibody

SouthernBiotech apoptosis related antibodies offer high sensitivity and low background for use in a variety of applications.

Specifications

Antigen Smac/DIABLO
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D12
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
Host Species Mouse
Immunogen Recombinant Smac/DIABLO
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 2-8°C
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.