Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMAD1/5/8/9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$207.00 - $499.50
Specifications
| Antigen | SMAD1/5/8/9 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056656T
![]() |
Novus Biologicals
NB10056656SS |
0.025 mg |
Each for $207.00
|
|
|||||
NB10056656
![]() |
Novus Biologicals
NB10056656 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
SMAD1/5/8/9 Polyclonal specifically detects SMAD1/5/8/9 in Human, Mouse samples. It is validated for Western Blot.Specifications
| SMAD1/5/8/9 | |
| Western Blot 1 ug/ml | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse | |
| Q15797 | |
| 4086 | |
| This antibody was developed against two synthetic peptides corresponding to amino acids 19-33 (GWKQGDEEEKWAEKA) and amino acids 315-329 (SNVNRNSTIENTRRH) of human SMAD1. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Diabetes Research | |
| PBS with 0.05% Sodium Azide | |
| BSP1, BSP-1, hSMAD1, JV4-1SMAD, mothers against DPP homolog 1 (Drosophila), MAD homolog 1, MADH1JV41, MADR1MAD, mothers against decapentaplegic homolog 1 (Drosophila), Mad-related protein 1, mothers against decapentaplegic homolog 1, Mothers against DPP homolog 1, SMAD family member 1SMAD 1, SMAD, mothers against DPP homolog 1, Smad1, TGF-beta signaling protein 1, transforming growth factor-beta signaling protein 1 | |
| SMAD1 | |
| IgG | |
| Protein G purified | |
| The synthetic peptide immunogens used to make this antibody are 100% conserved between SMAD1/5/8/9. Therefore, the antibody recognizes SMADs 1, 5, 8, and 9. The antibody may cross-react with SMAD2 or 3. The actual SMADs detected with this antibody may vary according to cell or tissue-type specific expression. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title