Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMAD1/5/8/9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | SMAD1/5/8/9 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5 - 2 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB600962SS
![]() |
Novus Biologicals
NB600962SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB600962
![]() |
Novus Biologicals
NB600962 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
SMAD1/5/8/9 Polyclonal specifically detects SMAD1/5/8/9 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SMAD1/5/8/9 | |
Western Blot 0.5 - 2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
0.1 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
4086 | |
This antibody was raised against two synthetic peptides corresponding to amino acids 18-32 (LGWKQGDEEEKWAEK) and amino acids 315-329 (SNVNRNSTIENTRRH) of human SMAD1. GenBank Accession No. AAP36050.1. They are 100% conserved or highly conserved between human and multiple species. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
BSP1, BSP-1, hSMAD1, JV4-1SMAD, mothers against DPP homolog 1 (Drosophila), MAD homolog 1, MADH1JV41, MADR1MAD, mothers against decapentaplegic homolog 1 (Drosophila), Mad-related protein 1, mothers against decapentaplegic homolog 1, Mothers against DPP homolog 1, SMAD family member 1SMAD 1, SMAD, mothers against DPP homolog 1, Smad1, TGF-beta signaling protein 1, transforming growth factor-beta signaling protein 1 | |
SMAD1 | |
IgG | |
Protein G purified | |
The amino acid sequences used to make this antibody are 100% conserved between SMAD1/5/8/9. Therefore, the antibody recognizes SMADs 1, 5, 8, and 9. The antibody may cross-react with SMAD2 or 3. The actual SMADs detected with this antibody may vary according to cell or tissue-type specific expression. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title