Learn More
SMAD1 Mouse anti-Human, Clone: OTI4H4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809093

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene ′mothers against decapentaplegic′ (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein mediates the signals of the bone morphogenetic proteins (BMPs), which are involved in a range of biological activities including cell growth, apoptosis, morphogenesis, development and immune responses. In response to BMP ligands, this protein can be phosphorylated and activated by the BMP receptor kinase. The phosphorylated form of this protein forms a complex with SMAD4, which is important for its function in the transcription regulation. This protein is a target for SMAD-specific E3 ubiquitin ligases, such as SMURF1 and SMURF2, and undergoes ubiquitination and proteasome-mediated degradation. Alternatively spliced transcript variants encoding the same protein have been observed.Specifications
SMAD1 | |
Monoclonal | |
Unconjugated | |
SMAD1 | |
BSP-1, BSP1, JV4-1, JV41, MADH1, MADR1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4086 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4H4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15797 | |
SMAD1 | |
Full length human recombit protein of human SMAD1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.