Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMAD1/SMAD5/SMAD9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104523
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104523 Supplier Invitrogen™ Supplier No. PA5104523
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Smad1/5/9.

The protein encoded by SMAD1/SMAD5/SMAD8/SMAD9 belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene 'mothers against decapentaplegic' (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein mediates the signals of the bone morphogenetic proteins (BMPs), which are involved in a range of biological activities including cell growth, apoptosis, morphogenesis, development and immune responses. In response to BMP ligands, this protein can be phosphorylated and activated by the BMP receptor kinase. The phosphorylated form of this protein forms a complex with SMAD4, which is important for its function in the transcription regulation. This protein is a target for SMAD-specific E3 ubiquitin ligases, such as SMURF1 and SMURF2, and undergoes ubiquitination and proteasome-mediated degradation. Alternatively spliced transcript variants encoding the same protein have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMAD1/SMAD5/SMAD9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SMAD9
Gene Accession No. O15198, O54835, P70340, P97454, P97588, Q15797, Q99717, Q9JIW5, Q9R1V3
Gene Alias 1110051M15Rik; AI451355; AI528653; BSP1; BSP-1; dwarfin-A; Dwarfin-C; dwf-A; DWFC; Dwf-C; JV41; JV4-1; JV5-1; MAD (mothers against decapentaplegic, Drosophila) homolog 1; MAD homolog 1; MAD homolog 5; MAD homolog 9; MAD homolog1 (mothers against decapentaplegic, Drosophila); MAD, mothers against decapentaplegic homolog 1; MAD, mothers against decapentaplegic homolog 5; MADH1; MADH5; MADH6; Madh8; Madh9; Madr1; mad-related protein 1; Mlp1; mMad1; mothers against decapentaplegic homolog 1; Mothers against decapentaplegic homolog 5; mothers against decapentaplegic homolog 9; mothers against decapentaplegic, drosophila, homolog of, 5; Mothers against decapentaplegic, drosophila, homolog of, 9; mothers against DPP homolog 1; mothers against DPP homolog 5; mothers against DPP homolog 9; mothers-against-DPP-related 1; Msmad5; MusMLP; PPH2; SMA- and MAD-related protein 5; SMAD; Smad 1; SMAD 5; SMAD 9; SMAD family member 1; SMAD family member 5; SMAD family member 9; SMAD, mothers against DPP homolog 1; SMAD, mothers against DPP homolog 5; SMAD, mothers against DPP homolog 9; SMAD1; Smad5; Smad8; SMAD8/9; SMAD8A; SMAD8B; Smad9; TGF-beta signaling protein 1; transforming growth factor-beta signaling protein 1; Transforming growth factor-beta-signaling protein 1
Gene Symbols SMAD1, Smad5, SMAD9
Host Species Rabbit
Immunogen A synthesized peptide derived from human SMAD1(Accession Q15797), corresponding to amino acid residues I438-S465.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17125, 17129, 25671, 4086, 4090, 4093, 55994, 59328, 85435
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.