Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smad2/3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$159.60 - $391.00
Specifications
Antigen | Smad2/3 |
---|---|
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Smad2/3 Polyclonal antibody specifically detects Smad2/3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Smad2/3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Cell Cycle and Replication, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
PBS, 50% glycerol, pH7.3 | |
4087 | |
IgG | |
Affinity purified |
Western Blot 1:500 - 1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
hMAD-2, hSMAD2, JV18, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2MGC34440, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2 | |
A synthetic phosphorylated peptide around S423 & S425 of human SMAD2/SMAD3SMAD3 (NP_005893.1). SSVS | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title