Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smad2, p Ser250 Antibody (4D12), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Smad2 |
---|---|
Clone | 4D12 |
Dilution | Western Blot 1:500-1:5000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
Smad2 Monoclonal antibody specifically detects Smad2 in Human samples. It is validated for Western Blot, ELISASpecifications
Smad2 | |
Western Blot 1:500-1:5000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
hMAD-2, hSMAD2, JV18, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2MGC34440, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2 | |
A synthesized peptide derived from Human SMAD2 [p Ser250] [UniProt Q15796] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
4D12 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Cancer, Cell Cycle and Replication, Metastasis, Phospho Specific, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
4087 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title