Learn More
Smad3 Antibody (378611), Alexa Fluor™ 350, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB4038U
Description
Smad3 Monoclonal antibody specifically detects Smad3 in Human samples. It is validated for Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
Smad3 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
378611 | |
Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
DKFZp586N0721, DKFZp686J10186, hMAD-3, hSMAD3, HsT17436, JV15-2MGC60396, MAD homolog 3, mad protein homolog, MAD, mothers against decapentaplegic homolog 3, MAD, mothers against decapentaplegic homolog 3 (Drosophila), mad3, MADH3mad homolog JV15-2, mothers against decapentaplegic homolog 3, Mothers against DPP homolog 3, SMA- and MAD-related protein 3, SMAD 3, SMAD family member 3HSPC193, SMAD, mothers against DPP homolog 3, SMAD, mothers against DPP homolog 3 (Drosophila), Smad3 | |
E. coli-derived recombinant human Smad3, Ser2-Ala230, Accession # P84022 | |
0.1 mL | |
Phospho Specific, Signal Transduction, Wnt Signaling Pathway | |
4088 | |
Store at 4C in the dark. | |
IgG1 |