Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smad3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
$206.50 - $427.50
Specifications
Antigen | Smad3 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 ug/ml |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056479T
![]() |
Novus Biologicals
NB10056479SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056479
![]() |
Novus Biologicals
NB10056479 |
0.1 mg |
Each for $427.50
|
|
|||||
Description
Smad3 Polyclonal specifically detects Smad3 in Human, Mouse, Rat, Porcine, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Smad3 | |
Western Blot 0.5-2 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Pig, Bovine, Canine, Primate | |
NP_005893 | |
4088 | |
A portion of amino acids 100-150 of human SMAD3 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Wnt Signaling Pathway | |
PBS with 0.05% Sodium Azide | |
DKFZp586N0721, DKFZp686J10186, hMAD-3, hSMAD3, HsT17436, JV15-2MGC60396, MAD homolog 3, mad protein homolog, MAD, mothers against decapentaplegic homolog 3, MAD, mothers against decapentaplegic homolog 3 (Drosophila), mad3, MADH3mad homolog JV15-2, mothers against decapentaplegic homolog 3, Mothers against DPP homolog 3, SMA- and MAD-related protein 3, SMAD 3, SMAD family member 3HSPC193, SMAD, mothers against DPP homolog 3, SMAD, mothers against DPP homolog 3 (Drosophila), Smad3 | |
SMAD3 | |
IgG | |
Protein G purified | |
Cross reacts with Bovine, Dog, Human, Mouse, Pig, Rat and Rhesus Monkey. Predicted to react with Zebrafish, Xenopus and Chicken. The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, dog, pig, bovine and rhesus monkey, and 90% homologous in chicken, xenopus and zebrafish. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title