Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smad3 Mouse anti-Human, Clone: PCRP-SMAD3-1A2, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Smad3 |
---|---|
Clone | PCRP-SMAD3-1A2 |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array |
Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
Description
Smad3 Monoclonal specifically detects Smad3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
Smad3 | |
0.2 mg/ml | |
Flow Cytometry, Immunofluorescence, Peptide Array | |
Unconjugated | |
Mouse | |
Phospho Specific, Signal Transduction, Wnt Signaling Pathway | |
10 mM PBS with 0.05% BSA | |
4088 | |
IgG1 | |
Protein A or G purified |
PCRP-SMAD3-1A2 | |
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DKFZp586N0721, DKFZp686J10186, hMAD-3, hSMAD3, HsT17436, JV15-2MGC60396, MAD homolog 3, mad protein homolog, MAD, mothers against decapentaplegic homolog 3, MAD, mothers against decapentaplegic homolog 3 (Drosophila), mad3, MADH3mad homolog JV15-2, mothers against decapentaplegic homolog 3, Mothers against DPP homolog 3, SMA- and MAD-related protein 3, SMAD 3, SMAD family member 3HSPC193, SMAD, mothers against DPP homolog 3, SMAD, mothers against DPP homolog 3 (Drosophila), Smad3 | |
Recombinant full-length human Smad3 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title