Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smad4 Antibody (SMAD4/2524), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308345AF594
Description
Smad4 Monoclonal antibody specifically detects Smad4 in Human samples. It is validated for ELISASpecifications
Smad4 | |
Monoclonal | |
Alexa Fluor 594 | |
DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4 | |
Recombinant full-length human Smad4 protein (Uniprot: Q13485 ) | |
100 μg | |
Signal Transduction, Stem Cell Signaling Pathway | |
4089 | |
Store at 4°C in the dark. | |
IgG2a κ |
ELISA | |
SMAD4/2524 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction