Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smad5, p Ser463, p Ser465 Antibody (2G11), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Smad5 |
---|---|
Clone | 2G11 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Smad5 Monoclonal antibody specifically detects Smad5 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Smad5 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
hSmad5, JV5-1DKFZp781O1323, MAD homolog 5, MAD, mothers against decapentaplegic homolog 5 (Drosophila), MADH5mothers against decapentaplegic homolog 5, mothers against decapentaplegic homolog 5, mothers against decapentaplegic, drosophila, homolog of, 5, Mothers against DPP homolog 5, SMA- and MAD-related protein 5, SMAD 5, SMAD family member 5DKFZp781C1895, SMAD, mothers against DPP homolog 5, SMAD, mothers against DPP homolog 5 (Drosophila), Smad5 | |
A synthesized peptide derived from Human SMAD5 [p Ser463, p Ser465] [UniProt Q99717] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
2G11 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Phospho Specific, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
4090 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title