Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMAD5 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: AFB-19, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20575
Description
SMAD5 Monoclonal antibody specifically detects SMAD5 in Human, Mouse, Rat samples. It is validated for ICC, WB-Ce, IHC-P.Specifications
SMAD5 | |
Monoclonal | |
Unconjugated | |
SMAD5 | |
Rabbit | |
100 μg | |
4090 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
AFB-19 | |
SMAD family member 5 | |
DKFZp781C1895, DKFZp781O1323, Dwfc, JV5-1, MADH5 | |
A synthetic phosphopeptide corresponding to residues surrounding of human SMAD5 | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction