Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMAD6 Antibody (5H3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP151651
Description
SMAD6 Monoclonal specifically detects SMAD6 in Human samples. It is validated for Western Blot, ELISA.Specifications
SMAD6 | |
Monoclonal | |
Unconjugated | |
hSMAD6, HsT17432, MAD homolog 6, MADH6SMAD, mothers against DPP homolog 6 (Drosophila), MADH7, mothers against decapentaplegic homolog 6, Mothers against decapentaplegic, drosophila, homolog of, 6, Mothers against DPP homolog 6, SMAD 6, SMAD family member 6MAD, mothers against decapentaplegic homolog 6 (Drosophila), SMAD, mothers against DPP homolog 6, Smad6 | |
Mouse | |
53 kDa | |
0.1 mL | |
Stem Cell Signaling Pathway | |
4091 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA | |
5H3 | |
Western Blot 1:500-1:2000, ELISA 1:10000 | |
SMAD6 | |
Purified recombinant fragment of human SMAD6 expressed in E. Coli. | |
Unpurified | |
RUO | |
Primary | |
Human | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction