Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smad7 Antibody - Azide Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB10056688
Description
Smad7 Polyclonal specifically detects Smad7 in Human, Mouse, Sheep samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
Smad7 | |
Polyclonal | |
Unconjugated | |
Sterile - filtered PBS with No Preservative | |
CRCS3, FLJ16482, hSMAD7, MAD homolog 7, MAD homolog 8, MAD, mothers against decapentaplegic homolog 7 (Drosophila), MADH7mothers against decapentaplegic homolog 7, MADH8, Mothers against decapentaplegic homolog 8, Mothers against decapentaplegic, drosophila, homolog of, 7, Mothers against DPP homolog 7, Mothers against DPP homolog 8, SMAD 7, SMAD family member 7MAD (mothers against decapentaplegic, Drosophila) homolog 7, SMAD, mothers against DPP homolog 7, SMAD, mothers against DPP homolog 7 (Drosophila), Smad7 | |
Rabbit | |
Protein G purified | |
RUO | |
4092 | |
Human, Mouse, Sheep | |
Purified |
Western Blot, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 4 - 6 ug/ml | |
NP_005895 | |
SMAD7 | |
This antibody was raised against two peptides made from within amino acids 10 and 50 of human SMAD7. This sequence is 100% or highly conserved between human and multiple species. | |
0.1 mg | |
Primary | |
In A549, an approx. 45 kDa band is observed. This may cross-react with SMAD6 depending on cell or tissue-specific expression. Cross reacts with Human, Mouse and Rat. Predicted to react with Chimpanzee, Dog, Pufferfish, Zebrafish and Xenopus. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction