Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smad9 Antibody (PCRP-SMAD9-2F4), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314117MFV500
Description
Smad9 Monoclonal antibody specifically detects Smad9 in Human samples. It is validated for Flow CytometrySpecifications
Smad9 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
4093 | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry | |
PCRP-SMAD9-2F4 | |
Flow Cytometry | |
MAD homolog 9, MAD, mothers against decapentaplegic homolog 9 (Drosophila), Madh6, MADH6mothers against decapentaplegic homolog 9, MADH9Mothers against decapentaplegic, drosophila, homolog of, 9, Mothers against DPP homolog 9, SMAD 9, SMAD family member 9SMAD8B, SMAD, mothers against DPP homolog 9, SMAD, mothers against DPP homolog 9 (Drosophila), SMAD8, SMAD8A, Smad9 | |
Recombinant full-length human Smad9 protein (Uniprot: O15198) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction