Learn More
Invitrogen™ SMAD9 Monoclonal Antibody (3H2)

Description
Peptide Sequence: MHSTTPISSL FSFTSPAVKR LLGWKQGDEE EKWAEKAVDS LVKKLKKKKG AMDELERALS CPGQPSKCVT IPRSLDGRLQ VSHCKGLPHV IYCRVWRWPD LQSHHELKPL ECCEFPFGSK QKEVCINPYH YRRVETPVLP PVLVPRHSEY NPQLSLLAKF RSASLHSEPL MPHNATYPDS FQQPPCSALP PSPSHAFSQS PCTASYPHSP GSPSEPESPY QHSDFRPVCY EEPQHWCSVA YYELNNRVGE TFQASSRSVL IDGFTDPSNN RNRFCLGLLS NVNRNSTIEN TRRHIGKGVH LYYVGGEVYA ECVSDSSIFV QSRNCNYQHG FHPATVCKIP SGCSLKVFNN QLFAQLLAQS VHHGFEVVYE LTKMCTIRMS FVKGWGAEYH RQDVTSTPCW IEIHLHGPLQ WLDKVLTQMG SPHNPISSVS.
Specifications
Specifications
| Antigen | SMAD9 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3H2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SMAD9 |
| Gene Accession No. | O15198 |
| Gene Alias | HGNC:6774; MAD; MAD homolog 9; MADH6; Madh8; MADH9; Mothers against decapentaplegic homolog 9; Mothers against decapentaplegic, drosophila, homolog of, 9; mothers against DPP homolog 9; PPH2; RP11-421P11.9; SMAD; SMAD 9; SMAD family member 9; SMAD, mothers against DPP homolog 9; SMAD8; SMAD8/9; SMAD8A; SMAD8B; SMAD9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.