Learn More
Invitrogen™ SMAD9 Polyclonal Antibody

Description
PA1-29826 has been successfully used in Western blot applications, where a band of 51 kDa is detected. A549 cells can be used as a positive control for this antibody.
Specifications
Specifications
| Antigen | SMAD9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | SMAD9 |
| Gene Accession No. | O15198 |
| Gene Alias | HGNC:6774; MAD; MAD homolog 9; MADH6; Madh8; MADH9; Mothers against decapentaplegic homolog 9; Mothers against decapentaplegic, drosophila, homolog of, 9; mothers against DPP homolog 9; PPH2; RP11-421P11.9; SMAD; SMAD 9; SMAD family member 9; SMAD, mothers against DPP homolog 9; SMAD8; SMAD8/9; SMAD8A; SMAD8B; SMAD9 |
| Gene Symbols | SMAD9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.