Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMAGP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558895 Supplier Invitrogen™ Supplier No. PA558895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YKNKGSYVTY EPTEGEPSAI VQMESDLAKG SEKEEYFI Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

Small cell adhesion glycoprotein (SMAGP) is a gene located on chromosome 1p33 that encodes a transmembrane protein involved in cell adhesion processes. SMAGP is primarily expressed in epithelial tissues and plays a role in maintaining cellular architecture and tissue integrity. This glycoprotein contributes to the organization of cell-cell junctions, which are critical for epithelial barrier function and communication between cells. Altered expression of SMAGP has been observed in various cancers, including gastric and colorectal cancers, where it may influence tumor progression and metastasis by affecting cellular adhesion and epithelial-to-mesenchymal transition (EMT). Research into SMAGP continues to explore its functional mechanisms in epithelial biology and its potential as a biomarker for cancer diagnosis and prognosis. By providing insights into the molecular pathways governing cell adhesion and tumorigenesis, studies on SMAGP contribute to the broader understanding of cancer biology and therapeutic opportunities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMAGP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMAGP
Gene Accession No. Q0VAQ4
Gene Alias hSMAGP; Smagp; Small cell adhesion glycoprotein; small cell transmembrane and glycosylated protein; small transmembrane and glycosylated protein; small trans-membrane and glycosylated protein
Gene Symbols SMAGP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMAGP. Recombinant protein control fragment (Product #RP-97826).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57228
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.