Learn More
Promega SmaI, 5,000u
Catalog No. PRR6125 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5,000 units
Catalog No. Quantity
PRR6125 5,000 units
1 options
Catalog No. PRR6125 Supplier Promega Supplier No. R6125
CCC/GGG GGG\CCC
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications
  • Available at high concentration, containing 5,000 units of SmaI at a concentration of 40−80u/microl
Specifications
Quantity 5,000 units