Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smallpox B5R Antibody (1J2F3), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP224699MFV500
Description
Smallpox B5R Monoclonal antibody specifically detects Smallpox B5R in Virus samples. It is validated for Western BlotSpecifications
Smallpox B5R | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
1486563 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot | |
1J2F3 | |
Western Blot | |
B5R protein, B6R, B7R, B7R protein, EEV membrane glycoprotein, Glycoprotein, H7R, H7R protein, ORF5R, VARVgp175 | |
Amino acids 33-47 (IQEYNLIRTYKKVDK) ofthe B5R protein were used as the immunogen (NCBI Reference Sequence: NP_042218.1) | |
0.1 mL | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction