Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smallpox B5R Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | Smallpox B5R |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.1-1 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056453T
![]() |
Novus Biologicals
NB10056453SS |
0.025 mg |
Each for $181.90
|
|
|||||
NB10056453
![]() |
Novus Biologicals
NB10056453 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
Smallpox B5R Polyclonal specifically detects Smallpox B5R in Virus samples. It is validated for Western Blot.Specifications
Smallpox B5R | |
Western Blot 0.1-1 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
Amino acids 200-214 (GSDVDKKDTHGNTPF) of the B5R protein were used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Virus | |
B5R protein, B6R, B7R, B7R protein, EEV membrane glycoprotein, Glycoprotein, H7R, H7R protein, ORF5R, VARVgp175 | |
IgG | |
Protein G purified | |
Reacts with Variola virus. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title