Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smallpox B5R Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen Smallpox B5R
Concentration 0.5 mg/mL
Dilution Western Blot 0.1-1 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056454T
SDP
View Documents
Novus Biologicals
NB10056454SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NB10056454
SDP
View Documents
Novus Biologicals
NB10056454
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

Smallpox B5R Polyclonal specifically detects Smallpox B5R in Virus samples. It is validated for Western Blot.
Specifications

Specifications

Smallpox B5R
Western Blot 0.1-1 ug/ml
Polyclonal
Purified
RUO
PBS containing 0.2% gelatin with 0.05% Sodium Azide
Amino acids 180-194 (HYKKSESESYNELIK) of the B5R protein were used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Virus
B5R protein, B6R, B7R, B7R protein, EEV membrane glycoprotein, Glycoprotein, H7R, H7R protein, ORF5R, VARVgp175
IgG
Protein G purified
Reacts with Varioa virus.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.