Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Smallpox Virus A27L Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144503
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144503 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144503 Supplier Invitrogen™ Supplier No. PA5144503
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Smallpox Virus A27L Polyclonal Antibody for Western Blot, ELISA

Smallpox Virus A27L is a structural protein involved in the envelopment of mature virion (MV) to form the wrapped virion (WV). The wrapping consists of the addition of Golgi membranes to the mature virion. Participates in mature virion (MV) movement within the infected cell. May play an indirect role in MV-cell fusion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Smallpox Virus A27L
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias VARVgp134
Host Species Rabbit
Immunogen Recombinant Vaccinia virus 14 kDa fusion protein (A27L) (1-110AA).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.