Learn More
SMAP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304687AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
C11orf58 is a protein coding gene.Specifications
SMAP | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O00193 | |
C11orf58 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
C11orf58 | |
IMAGE145052, SMAP, small acidic protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10944 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.