Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110397
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110397 Supplier Invitrogen™ Supplier No. PA5110397
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPEKPAKPLT AEKLQKKDQQ LEPKKSTSPK KAAEPTVDLL GLDGPAVAPV TNGNTTVPPL NDDLDIFGPM I.

The protein encoded by this gene is similar to the mouse stromal membrane-associated protein-1. This similarity suggests that this human gene product is also a type II membrane glycoprotein involved in the erythropoietic stimulatory activity of stromal cells. Alternate splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMAP1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SMAP1
Gene Accession No. Q8IYB5, Q91VZ6
Gene Alias 1700056O10Rik; 4921514B13Rik; 4921525H11Rik; AI462175; small ArfGAP 1; SMAP1; SMAP-1; stromal membrane-associated GTPase-activating protein 1; stromal membrane-associated protein 1
Gene Symbols SMAP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 180-250 of human SMAP1 (NP_001037770).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60682, 684800, 98366
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.