Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556509
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556509 Supplier Invitrogen™ Supplier No. PA556509
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KAAEPTVDLL GLDGPAVAPV TNGNTTVPPL NDDLDIFGPM ISNPLPATVM PPAQGTPSAP AAATLSTVTS GDLDLFTEQT TKSEEVAKKQ LSKDSIL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

The protein encoded by this gene is similar to the mouse stromal membrane-associated protein-1. This similarity suggests that this human gene product is also a type II membrane glycoprotein involved in the erythropoietic stimulatory activity of stromal cells. Alternate splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMAP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMAP1
Gene Accession No. Q8IYB5, Q91VZ6
Gene Alias 1700056O10Rik; 4921514B13Rik; 4921525H11Rik; AI462175; small ArfGAP 1; SMAP1; SMAP-1; stromal membrane-associated GTPase-activating protein 1; stromal membrane-associated protein 1
Gene Symbols SMAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMAP1. Recombinant protein control fragment (Product #RP-94684).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60682, 684800, 98366
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.