Learn More
Invitrogen™ SMAP2 Monoclonal Antibody (OTI6E11)

Description
SMAP2 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | SMAP2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI6E11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Smap2 |
| Gene Accession No. | Q8WU79 |
| Gene Alias | 1810031K02Rik; AA408614; RGD1308418; small ArfGAP 2; small ArfGAP2; SMAP1L; SMAP2; stromal membrane-associated GTPase-activating protein 2; Stromal membrane-associated protein 1-like; Stromal membrane-associated protein 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.