Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554416 Supplier Invitrogen™ Supplier No. PA554416
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASGMVAPMAM PAGYMGGMQA SMMGVPNGMM TTQQAGYMAG MAAMPQTVYG VQPAQQLQWN LTQMTQQMAG MNFYGANGMM NYGQSMSGGN GQAANQTLSP QMWK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

SMAP2 (Small ArfGAP2) is a Protein Coding gene. Among its related pathways are Endocytosis. Gene Ontology (GO) annotations related to this gene include GTPase activator activity. An important paralog of this gene is SMAP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMAP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Smap2
Gene Accession No. Q7TN29, Q8WU79
Gene Alias 1810031K02Rik; AA408614; RGD1308418; small ArfGAP 2; small ArfGAP2; SMAP1L; SMAP2; stromal membrane-associated GTPase-activating protein 2; Stromal membrane-associated protein 1-like; Stromal membrane-associated protein 2
Gene Symbols Smap2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMAP2. Recombinant protein control fragment (Product #RP-93122).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298500, 64744, 69780
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.