Learn More
SMAP2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305772AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
SMAP2 (Small ArfGAP2) is a Protein Coding gene. Among its related pathways are Endocytosis. Gene Ontology (GO) annotations related to this gene include GTPase activator activity. An important paralog of this gene is SMAP1.Specifications
SMAP2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q7TN29, Q8WU79 | |
SMAP2 | |
between 379 and 429 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SMAP2 | |
SMAP1L, stromal membrane-associated GTPase-activating protein 2, stromal membrane-associated protein 1-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64744, 69780 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.