Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMARCA1 Monoclonal Antibody (2H7B8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531911
Description
MA5-31911 has been tested in indirect ELISA.
Members of this G protein family are characterized by the presence of conserved SNF2 domain, a helicase-like region. They also have an ATPase activity and are thought to have a significant role regulating chromatin remodeling. Although SNF2L1 has a ubiquitous expression pattern, its active form is expressed in neurons suggesting a possible role in promoting neurite outgrowth and regulating neuronal physiology. Principal Names: SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 1 (SMARCA1); SNF2L1; SNF2LB; Possible global transcription activator SNF2L1; SWI2 Official Gene Symbol: SMARCA1 Gen Bank Accession Number: NP_620604 Gene ID: 6594 (Human) Gene Map Locus: Xq25 (Human) SNF2L1, a ~123KDa protein, is a member of SWI/SNF family of proteins.
Specifications
SMARCA1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P28370, Q6PGB8 | |
Smarca1 | |
Purified recombinant fragment of human SMARCA1 (AA: 933-1070) expressed in E. coli. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2H7B8 | |
Unconjugated | |
Smarca1 | |
5730494M04Rik; ATP-dependent helicase SMARCA1; DKFZp686D1623; DNA-dependent ATPase SNF2L; FLJ41547; global transcription activator homologous sequence; hSNF2L; ISWI; nucleosome-remodeling factor subunit SNF2L; NURF140; Probable global transcription activator SNF2L1; RGD1561046; RP3-353H6.2; SMARCA1; SNF2L; SNF2L1; SNF2LB; SNF2-like 1; SNF2L-like protein; SNF2LT; sucrose nonfermenting 2-like protein 1; SWI; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1; SWI2 | |
Mouse | |
Protein G | |
RUO | |
6594, 93761 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction