Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCA1 Mouse anti-Human, Mouse, Clone:2H7B8, Abnova™

Catalog No. 89333904 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-904 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-904 Supplier Abnova Corporation Supplier No. MAB17517
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human SMARCA1.

This gene encodes a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen SMARCA1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2H7B8
Conjugate Unconjugated
Description SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry
Flow Cytometry (1:200-1:400)
Immunohistochemistry (1:200-1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene SMARCA1
Gene Alias DKFZp686D1623, FLJ41547, ISWI, NURF140, SNF2L, SNF2L1, SNF2LB, SWI, SWI2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 933-1070 of human SMARCA1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 6594
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.