Learn More
Invitrogen™ SMARCA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587744
Description
Immunogen sequence: MEQDTAAVAA TVAAADATAT IVVIEDEQPG PSTSQEEGAA AAATEATAAT EKGEKKKEKN VSSFQLKLAA KAPKSEKEMD PEYEEKMKAD RAKRFEFLLK QTELFAHFIQ PSAQKSPTSP LNMKLGRPRI KKDEKQSLIS AGDYRHRRTE QEEDEELLSE SRKTSNVCIR FEVSPSYVKG GPLRDYQIRG LNWLISLYEN; Positive Samples: HeLa, 293T, A-549, HT-1080, LO2; Cellular Location: Nucleus.
Members of this G protein family are characterized by the presence of conserved SNF2 domain, a helicase-like region. They also have an ATPase activity and are thought to have a significant role regulating chromatin remodeling. Although SNF2L1 has a ubiquitous expression pattern, its active form is expressed in neurons suggesting a possible role in promoting neurite outgrowth and regulating neuronal physiology. Principal Names: SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 1 (SMARCA1); SNF2L1; SNF2LB; Possible global transcription activator SNF2L1; SWI2 Official Gene Symbol: SMARCA1 Gen Bank Accession Number: NP_620604 Gene ID: 6594 (Human) Gene Map Locus: Xq25 (Human) SNF2L1, a ~123KDa protein, is a member of SWI/SNF family of proteins.
Specifications
SMARCA1 | |
Polyclonal | |
Unconjugated | |
Smarca1 | |
5730494M04Rik; ATP-dependent helicase SMARCA1; DKFZp686D1623; DNA-dependent ATPase SNF2L; FLJ41547; global transcription activator homologous sequence; hSNF2L; ISWI; nucleosome-remodeling factor subunit SNF2L; NURF140; Probable global transcription activator SNF2L1; RGD1561046; RP3-353H6.2; SMARCA1; SNF2L; SNF2L1; SNF2LB; SNF2-like 1; SNF2L-like protein; SNF2LT; sucrose nonfermenting 2-like protein 1; SWI; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1; SWI2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6594 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
2.40 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P28370 | |
Smarca1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human SMARCA1 (NP_0030602). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.