Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMARCA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111587
Description
Immunogen sequence: KGEKKKEKNV SSFQLKLAAK APKSEKEMDP EYEEKMKADR AKRFEFLLKQ.
Members of this G protein family are characterized by the presence of conserved SNF2 domain, a helicase-like region. They also have an ATPase activity and are thought to have a significant role regulating chromatin remodeling. Although SNF2L1 has a ubiquitous expression pattern, its active form is expressed in neurons suggesting a possible role in promoting neurite outgrowth and regulating neuronal physiology. Principal Names: SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 1 (SMARCA1); SNF2L1; SNF2LB; Possible global transcription activator SNF2L1; SWI2 Official Gene Symbol: SMARCA1 Gen Bank Accession Number: NP_620604 Gene ID: 6594 (Human) Gene Map Locus: Xq25 (Human) SNF2L1, a ~123KDa protein, is a member of SWI/SNF family of proteins.
Specifications
| SMARCA1 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P28370 | |
| Smarca1 | |
| Recombinant Protein corresonding to Human SMARCA1. Recombinant protein control fragment (Product #RP-107362). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| Unconjugated | |
| Smarca1 | |
| 5730494M04Rik; ATP-dependent helicase SMARCA1; DKFZp686D1623; DNA-dependent ATPase SNF2L; FLJ41547; global transcription activator homologous sequence; hSNF2L; ISWI; nucleosome-remodeling factor subunit SNF2L; NURF140; Probable global transcription activator SNF2L1; RGD1561046; RP3-353H6.2; SMARCA1; SNF2L; SNF2L1; SNF2LB; SNF2-like 1; SNF2L-like protein; SNF2LT; sucrose nonfermenting 2-like protein 1; SWI; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1; SWI2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6594 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction