Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCA1/SNF2L Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571722
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1722 10 μL
50-157-1721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1722 Supplier Bethyl Laboratories, Inc Supplier No. IHC00230T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Horse, Panda, Orangutan, Monkey, Gorilla, Northern white-cheeked gibbon, Naked mole r

Principal Names: SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 1 (SMARCA1); SNF2L1; SNF2LB; Possible global transcription activator SNF2L1; SWI2 Official Gene Symbol: SMARCA1 Gen Bank Accession Number: NP_620604 Gene ID: 6594 (Human) Gene Map Locus: Xq25 (Human) SNF2L1, a ∽116KDa protein, is a member of SWI/SNF family of proteins. Members of this family are characterized by the presence of conserved SNF2 domain, a helicase-like region. They also have an ATPase activity and are thought to have a significant role in regulation of Chromatin remodeling. Although SNF2L1 has a ubiquitous expression pattern, its active form is expressed in neurons suggesting a possible role in promoting neutrite outgrowth and regulating neuronal physiology.

Specifications

Antigen SMARCA1/SNF2L
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SMARCA1
Gene Accession No. P28370
Gene Alias SWI/SNF related, matrix associated, actin dependent regulator of Chromatin, subfamily a member 1, DKFZp686D1623, FLJ41547, ISWI, NURF140, SNF2L, SNF2L1, SNF2LB, SWI, SWI2, SNF2LT
Gene Symbols SMARCA1
Host Species Rabbit
Immunogen Between 1004 and 1054
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6594
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.