Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMARCA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583119
Description
Immunogen sequence: SPVQLHQLRA QILAYKMLAR GQPLPETLQL AVQGKRTLPG LQQQQQQQQQ QQQQQQQQQQ QQQQPQQQPP QPQTQQQQQP ALVNYNRPSG PGPELSGPST PQKLPVP.
SMARCA2 encodes a protein that is a member of the SWI/SNF family of proteins and is highly similar to the brahma protein of Drosophila. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. Two transcript variants encoding different isoforms have been found for this gene, which contains a trinucleotide repeat (CAG) length polymorphism.
Specifications
SMARCA2 | |
Polyclonal | |
Unconjugated | |
SMARCA2 | |
2610209L14Rik; ATP-dependent helicase SMARCA2; BAF190; Baf190b; brahma; BRG1-associated factor 190B; BRM; FLJ36757; global transcription activator homologous sequence; hBRM; hSNF2a; MGC74511; NCBRS; Probable global transcription activator SNF2L2; protein brahma homolog; SMARCA2; SNF2; SNF2/SWI2-like protein 2; SNF2A; SNF2alpha; SNF2-alpha; Snf2l2; SNF2LA; Sth1p; subunit of the SWI/SNF chromatin remodeling complex; sucrose nonfermenting 2-like protein 2; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 2; SWI2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6595 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.12 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P51531 | |
SMARCA2 | |
Recombinant protein corresponding to Human SMARCA2. Recombinant protein control fragment (Product #RP-102384). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction