Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCA4 / BRG1 Monoclonal Antibody (3G4), Active Motif™
SDP

Catalog No. 501992871
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2871 100 μg
50-199-2872 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2871 Supplier Active Motif Supplier No. 39807
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Brg1 (Brahma-related gene 1) is an ATPase subunit of SWI2/SNF2-like chromatin-remodeling complexes that enable access of regulatory and effector proteins in transcription, DNA repair and DNA replication. Although Brg1-containing complexes are not essential for general cell survival, they participate in transcriptional regulation of several hundred genes including those involved in interferon and stress response, immune cells differentiation, neurogenesis, cell cycle etc. and is absolutely necessary for mouse embryogenesis. Brg1 is also involved in cell growth arrest, senescence and tumour supression.

Specifications

Antigen SMARCA4 / BRG1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene Smarca4
Gene Accession No. P51532, Q3TKT4
Gene Alias ATP-dependent helicase SMARCA4; b2b508.1Clo; b2b692Clo; BAF190; BAF190A; brahma protein-like 1; Brg1; brg-1; BRG1-associated factor 190A; BRM/SWI2-related gene 1; CSS4; FLJ39786; global transcription activator homologous sequence; homeotic gene regulator; HP1-BP72; hSNF2b; Mitotic growth and transcription activator; MRD16; nuclear protein GRB1; Protein brahma homolog 1; Protein BRG-1; RTPS2; SMARCA4; SNF2; SNF2B; SNF2beta; SNF2-beta; Snf2l4; SNF2LB; SNF2-like 4; sucrose nonfermenting-like 4; SW1/SNF; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; SWI2; Transcription activator BRG1
Gene Symbols Smarca4
Host Species Rat
Immunogen This SMARCA4 / BRG1 antibody was raised against a recombinant protein corresponding to amino acids 214-279 of mouse BRG1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20586, 6597
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.