Learn More
SMARCA4/BRG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
The antibody is specific to SMARCA4.
Specifications
Specifications
| Antigen | SMARCA4/BRG1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SMARCA4 |
| Gene Accession No. | P51532, Q3TKT4, Q8K1P7 |
| Gene Alias | ATP dependent helicase SMARCA4, BAF190, BAF190A, BRG1, BRG1 associated factor 190A, hSNF2b, Protein brahma homolog 1, Protein BRG 1, SMARCA4, SNF2, SNF2 BETA, SNF2B, SNF2L4, SNF2LB, SWI2, Transcription activator BRG1 |
| Gene Symbols | Smarca4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.